missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Sulfate, Pentahydrate, Granular, FCC, 98-102%, Spectrum™ Chemical
Description
Spectrum™ Chemical Copper Sulfate, Pentahydrate, Granular, FCC is used as a fungicide, algaecide and feed supplement. Find a high quality inventory of Copper Copper Sulfate, Pentahydrate, Granular, FCC. Approved by the Food Chemical Codex, it is used as a fungicide, algaecide, and feed supplement, and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| UN Number | UN3077 |
| Quantity | 125 g |
| Percent Purity | 98 to 102% |
| Grade | FCC |
| Packaging | Poly Bottle |