missing translation for 'onlineSavingsMsg'
Learn More

Copper Sulfate, Certified, 10.0% (w/v) ±0.2%, LabChem™

Catalog No. p-5388122
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC134401 500 mL
LC134402 1 L
LC134404 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC134401 Supplier LabChem Supplier No. LC134401
Only null left

Chemical Identifiers

CAS 7758-99-8 , 7732-18-5
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Color Blue
CAS Max % 90, 10
Physical Form Liquid
Quantity 500 mL
Linear Formula CuSO4·5H2O
Solubility Information Soluble in water
Formula Weight 249.68
Percent Purity 10.0% ±0.2% (w/v)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Sulfur compounds; Copper
Chemical Name or Material Copper Sulfate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature