missing translation for 'onlineSavingsMsg'
Learn More

Copper Sulfate, Certified, 10.0% (w/v) ±0.2%, LabChem™

Supplier:  LabChem LC134402

 View more versions of this product

Catalog No. LC134402


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Copper Sulfate
Blue
Liquid
CuH10O9S
MFCD00149681
Soluble in water
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
249.68
CHEBI:31440
10.0% ±0.2% (w/v)
Passes Test
Sulfur compounds; Copper
7758-99-8,7732-18-5
90,10
1 L
CuSO4·5H2O
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper(2+) pentahydrate sulfate
24463
249.68
Certified
Poly Bottle

Chemical Identifiers

7758-99-8
249.68
JZCCFEFSEZPSOG-UHFFFAOYSA-L
24463
copper(2+) pentahydrate sulfate
CuH10O9S
MFCD00149681
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
CHEBI:31440
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions