missing translation for 'onlineSavingsMsg'
Learn More

Copper Sulfate, Certified, 1.186 ±0.003, LabChem™

Catalog No. LC134452
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC134452 1 L
LC134454 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC134452 Supplier LabChem Supplier No. LC134452
Only null left

Chemical Identifiers

CAS 7758-99-8 , 7732-18-5
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Color Blue
CAS Max % 70.48, 29.52
Physical Form Liquid
Quantity 1 L
Linear Formula CuSO4·5H2O
Solubility Information Soluble in water
Formula Weight 249.68
Grade Certified
Concentration or Composition (by Analyte or Components) 1.186 ±0.003
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.186 at 18.4°C
Decomposition Information Sulfur compounds; Copper
Chemical Name or Material Copper Sulfate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Do not eat, drink or smoke when using this product.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Collect spillage.
Recommended Storage Room Temperature