missing translation for 'onlineSavingsMsg'
Learn More

Copper Sulfate, Certified, 0.0200M ±0.0002M (0.02N), LabChem™

Catalog No. LC134432
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC134432 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC134432 Supplier LabChem Supplier No. LC134432

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Standardized aqueous solution for laboratory applications.

Chemical Identifiers

CAS 7758-99-8 , 7732-18-5
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Color Blue
CAS Max % 99.5, 0.5
Physical Form Liquid
Quantity 1 L
Linear Formula CuSO4·5H2O
Solubility Information Soluble in water
Formula Weight 249.68
Grade Certified
Concentration or Composition (by Analyte or Components) 0.0200M ±0.0002M (0.02N)
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Sulfur compounds
Density 1g/mL
Chemical Name or Material Copper Sulfate
Show More Show Less
Hazard Statement GHS H Statement
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature