missing translation for 'onlineSavingsMsg'
Learn More

Copper Sulfate, 4% w/v, Ricca Chemical Company

Catalog No. R23188004B
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R23188004B 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R23188004B Supplier Ricca Chemical Company Supplier No. R23188004B

Chemical Identifiers

CAS 7758-99-8 , 7732-18-5

Specifications

Boiling Point ~100°C
Color Blue
Physical Form Liquid
Quantity 4 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 5% w/v
Chemical Name or Material Copper Sulfate Solution
ShelfLife 24 Months