Learn More
Copper, plasma standard solution, Specpure™ Cu 10,000μg/mL
$142.82 - $527.82
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA14372AD
|
Thermo Scientific Chemicals
014372AD |
50 mL |
Each for $142.82
|
|
|||||
|
AA14372AE
|
Thermo Scientific Chemicals
014372AE |
100 mL |
Each for $210.07
|
|
|||||
|
AA14372AP
|
Thermo Scientific Chemicals
014372AP |
500 mL |
Each for $527.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| MFCD00010965 |
Specifications
| Liquid | |
| MFCD00010965 | |
| 50 mL | |
| Copper plasma standard solution, Cu 10000μg/mL |
| Cu in 5% HNO3 | |
| UN3264 | |
| Specpure |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 231-714-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only