missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Oxide Wire, ACS Grade, 0.65 x 6mm, For Elementary Analysis, MilliporeSigma™
$341.89 - $758.14
Chemical Identifiers
| CAS | 1317-38-0 |
|---|---|
| Molecular Formula | CuO |
| Molecular Weight (g/mol) | 79.55 |
| MDL Number | MFCD00010979 |
| InChI Key | KKCXRELNMOYFLS-UHFFFAOYSA-N |
| Synonym | cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide |
| PubChem CID | 14829 |
| IUPAC Name | copper(2+) oxidandiide |
| SMILES | [O--].[Cu++] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
M1027670250
|
Technidata
1027670250 |
250 g | Glass bottle |
Each for $341.89
|
|
||||
|
M1027671000
|
Technidata
1027671000 |
1 kg | Glass bottle |
Each for $758.14
|
|
||||
Description
For elementary analysisChemical Identifiers
| 1317-38-0 | |
| 79.55 | |
| KKCXRELNMOYFLS-UHFFFAOYSA-N | |
| 14829 | |
| [O--].[Cu++] |
| CuO | |
| MFCD00010979 | |
| cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide | |
| copper(2+) oxidandiide |
Specifications
| 1317-38-0 | |
| 250 g | |
| MFCD00010979 | |
| cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide | |
| [O--].[Cu++] | |
| 79.55 | |
| 79.55 | |
| Odorless | |
| Dark gray wire (0.65 x 6mm) |
| Wire | |
| CuO | |
| UN 3077 | |
| KKCXRELNMOYFLS-UHFFFAOYSA-N | |
| copper(2+) oxidandiide | |
| 14829 | |
| ACS | |
| Glass bottle | |
| Copper oxide, Surface: CuO, Core: Cu2O |
Safety and Handling
H302: Harmful if swallowed.
H410: Very toxic to aquatic life with long lasting effects.
DOTInformation : DOT Class: 9, Packaging Group: III
EINECSNumber : 215-269-1