missing translation for 'onlineSavingsMsg'
Learn More

Copper Oxide Wire, ACS Grade, 0.65 x 6mm, For Elementary Analysis, MilliporeSigma™

Catalog No. M1027670250
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1027670250 250 g Glass bottle
M1027671000 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. M1027670250 missing translation for 'mfr' Technidata missing translation for 'supplierNo' 1.02767.0250
For elementary analysis

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
IUPAC Name copper(2+) oxidandiide
SMILES [O--].[Cu++]

Specifications

Physical Form Wire
Quantity 250 g
UN Number UN 3077
Formula Weight 79.55
Grade ACS
Odor Odorless
Packaging Glass bottle
Appearance Dark gray wire (0.65 x 6mm)
Chemical Name or Material Copper oxide, Surface: CuO, Core: Cu2O
missing translation for 'healthHazard1' H302: Harmful if swallowed.
H410: Very toxic to aquatic life with long lasting effects.
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 215-269-1