missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) Phthalocyanine (beta-form) 93.0+%, TCI America™
Specifications
Specifications
| Melting Point | 480°C |
| Color | Blue-Purple |
| Quantity | 25 g |
| Formula Weight | 576.08 |
| Percent Purity | ≥93.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Copper(II) Phthalocyanine (beta-form) |
Safety and Handling
| EINECSNumber | (5)-3299&(5)-5216 |
| RTECSNumber | GL8510000 |
| TSCA | Yes |