missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Phthalocyanine (beta-form) 93.0+%, TCI America™

Catalog No. P100625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P100625G 25 g
P1006100G 100 g
P1006500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P100625G Supplier TCI America Supplier No. P100625G

Chemical Identifiers

CAS 147-14-8
Molecular Formula C32H16CuN8
MDL Number MFCD00010719
Synonym Phthalocyanine Copper(II), CuPc

Specifications

Melting Point 480°C
Color Blue-Purple
Quantity 25 g
Formula Weight 576.08
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Copper(II) Phthalocyanine (beta-form)
EINECSNumber (5)-3299&(5)-5216
RTECSNumber GL8510000
TSCA Yes