missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) phosphate, 98%

Catalog No. aaa173600e
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA173600E 2500 g
AAA1736022 100 g
AAA1736036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA173600E Supplier Thermo Scientific Chemicals Supplier No. A173600E

CAS: 7798-23-4 | Cu3O8P2 | 380.578 g/mol

Copper(II) phosphate is used as organic catalyst, fertilizer, emulsifier, corrosion inhibitor for phosphoric acid, metal protectant, and animal feed additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) phosphate is used as organic catalyst, fertilizer, emulsifier, corrosion inhibitor for phosphoric acid, metal protectant, and animal feed additive.

Solubility
Insoluble with water and ethanol. Soluble in ammonia, ammonium hydroxide, acetone.

Notes
Hygroscopic. Incompatible with strong bases, metals, alkali metals and powdered metals.

Chemical Identifiers

CAS 7798-23-4
Molecular Formula Cu3O8P2
Molecular Weight (g/mol) 380.578
MDL Number MFCD00036268
InChI Key GQDHEYWVLBJKBA-UHFFFAOYSA-H
Synonym copper ii phosphate, cupric phosphate, unii-n8np6fr80r, tricopper bis orthophosphate, n8np6fr80r, copper phosphate cu3 po4 2, copper phosphate 3:2, hsdb 267, cupric phosphate cu3 po4 2, phosphoric acid, copper 2+ salt
PubChem CID 86469
IUPAC Name tricopper;diphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Cu+2].[Cu+2].[Cu+2]

Specifications

Melting Point >300°C
Quantity 2500 g
Linear Formula Cu3(PO4)2
Merck Index 14,2648
Solubility Information Insoluble with water and ethanol. Soluble in ammonia,ammonium hydroxide,acetone.
Formula Weight 380.58
Percent Purity 98%
Odor Odorless
Chemical Name or Material Copper(II) phosphate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 232-254-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only