accessibility menu, dialog, popup

UserName

CuCuCuOPOOOOPOOO

CAS RN 7798-23-4

IUPAC Name: tricopper(2+) diphosphate
Synonyms: copper(II) phosphate
Molecular Weight (g/mol): 380.58
Molecular Formula: Cu3O8P2
InChi Key: GQDHEYWVLBJKBA-UHFFFAOYSA-H
SMILES: [Cu++].[Cu++].[Cu++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results