missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Oxide Powder, Extra Pure, MilliporeSigma™

Catalog No. p-4939041
Change view
Click to view available options
Quantity:
500 g
25 kg
Packaging:
Plastic bottle
Plastic drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1027610500 500 g Plastic bottle
M1027619025 25 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1027610500 Supplier Technidata Supplier No. 1.02761.0500
Only null left

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
IUPAC Name copper(2+) oxidandiide
SMILES [O--].[Cu++]

Specifications

Physical Form Solid
Quantity 500 g
UN Number UN 3077
Formula Weight 79.55
Grade Extra Pure
Odor Odorless
Packaging Plastic bottle
Appearance Black solid
Chemical Name or Material Copper(II) Oxide Powder
Hazard Category H302: Harmful if swallowed.
H410: Very toxic to aquatic life with long lasting effects.
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 215-269-1