missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) oxide, nanopowder

Catalog No. p-7059468
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4466309 10 g
AA4466318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4466309 Supplier Thermo Scientific Chemicals Supplier No. 04466309
Only null left

CAS: 1317-38-0 | CuO | 79.55 g/mol

Copper(II) oxide nanopowder is used as a burning rate catalyst in rocket propellant, which enhances the the homogeneous propellant burning rate, lower pressure index. It can act as a catalyst used in superconducting materials, thermoelectric materials, sensing materials, glass and ceramics. It plays a vital role in photoconductive and photothermal applications. It is also used in ceramic resistors, magnetic storage media, gas sensors, near-infrared tilters, semiconductors, solar energy transformation and high-tech superconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
SMILES [O--].[Cu++]

Specifications

Melting Point 1326°C
Color Black
Physical Form Powder
Quantity 10 g
UN Number UN3077
Merck Index 14,2646
Solubility Information Practically insoluble in water,and alcohol. Soluble in dilute acids,alkali cyanides,and ammonium carbonate solution.
IUPAC Name copper(2+) oxidandiide
Formula Weight 79.55
Odor Odorless
Refractive Index 2.63
Density 6.3 g/mL
Chemical Name or Material Copper (II) oxide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 215-269-1
RTECSNumber GL7900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only