missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) oxide, Solstice

Catalog No. 6000449
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000449 100 g
6000450 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000449 Supplier Solstice Supplier No. 61200100G

Puriss. p.a.,≥99.0% (RT), powder

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
IUPAC Name oxocopper
SMILES [O--].[Cu++]

Specifications

Quantity 100 g
UN Number UN3077
Formula Weight 79.55g/mol
Percent Purity ≥99% (RT)
Chemical Name or Material Copper(II) oxide
Hazard Statement P273
RTECSNumber GL7900000