missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) oxide, 99+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 221280025

Catalog No. AC221280025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Copper(II) oxide
1317-38-0
99% min. (Complexometry)
CuO
04,108; 05,161
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
QPLDLSVMHZLSFG-UHFFFAOYSA-N
oxocopper
14829
99+%
p.a.
2.5kg
CuO
MFCD00010979
15, 2636
Solubility in water: insoluble. Other solubilities: soluble in dil.acids- slowly soluble in nh3,soluble in alkali cyanides and (nh4)2co3 solutions,practically insoluble in alcohol
O=[Cu]
79.54
79.54
Plastic bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

EINECSNumber : 215-269-1

RTECSNumber : GL7900000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only