missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) formate tetrahydrate, 98%

Catalog No. aaa1856930
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1856930 250 g
AAA1856918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1856930 Supplier Thermo Scientific Chemicals Supplier No. A1856930

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5893-61-8 | C2H10CuO8 | 225.64 g/mol

It is employed as an antibacterial agent in the treatment of cellulose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an antibacterial agent in the treatment of cellulose.

Solubility
Soluble in water (125 g/L at 20°C). Very slightly soluble in alcohol. Practically insoluble in most organic solvents.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 5893-61-8
Molecular Formula C2H10CuO8
Molecular Weight (g/mol) 225.64
MDL Number MFCD00167128
InChI Key LSIWWRSSSOYIMS-UHFFFAOYSA-L
Synonym copper ii formate tetrahydrate, cupric formate tetrahydrate, copper diformate tetrahydrate, unii-v7ra28204j, copper 2+ formate tetrahydrate, cupric formate tetrahydrate mi, copper formate cu hcoo 2 tetrahydrate, formic acid copper salt 2:1 tetrahydrate, acmc-209m8c
PubChem CID 6432244
IUPAC Name copper;diformate;tetrahydrate
SMILES C(=O)[O-].C(=O)[O-].O.O.O.O.[Cu+2]

Specifications

Quantity 250 g
UN Number UN3077
Merck Index 14,2639
Solubility Information Soluble in water (125g/L at 20°C). Very slightly soluble in alcohol. Practically insoluble in most organic solvents.
Formula Weight 225.65 (153.59 Anhydrous)
Percent Purity 98%
Chemical Name or Material Copper(II) formate tetrahydrate
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 208-865-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only