missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) ethoxide, 98%

Catalog No. p-7080058
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4631606 5 g
AA4631614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4631606 Supplier Thermo Scientific Chemicals Supplier No. 04631606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2850-65-9 | C4H10CuO2 | 153.668 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon

Chemical Identifiers

CAS 2850-65-9
Molecular Formula C4H10CuO2
Molecular Weight (g/mol) 153.668
MDL Number MFCD00015650
InChI Key CRCKGIUJMFFISH-UHFFFAOYSA-N
Synonym copper ii ethoxide, copper diethoxide, ethanol, copper 2 salt, copper ethoxide, cupric ethoxide, acmc-20aldp, copper 2+ bis ethoxide, copper 2+ ion bis ethoxide
PubChem CID 14419876
IUPAC Name copper;ethanolate
SMILES CC[O-].CC[O-].[Cu+2]

Specifications

Melting Point ∼120°C (decomposition)
Color Green
Linear Formula Cu(OC2H5)2
Quantity 5 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Formula Weight 153.67
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Copper (II) ethoxide
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
TSCA No
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures

RUO – Research Use Only