missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Disodium Ethylenediaminetetraacetate Tetrahydrate 98.0+%, TCI America™

$57.79 - $337.40

Chemical Identifiers

CAS 39208-15-6
Molecular Formula C10H20CuN2Na2O12
Molecular Weight (g/mol) 469.798
MDL Number MFCD00148920
InChI Key CHPMNDHAIUIBSK-UHFFFAOYSA-J
Synonym copper ii disodium ethylenediaminetetraacetate tetrahydrate, ethylenediaminetetraacetic acidcopper ii disodium salt tetrahydrate, acmc-209j3w, edta-cu-15, ksc911m0f, copper 2+ disodium ion 4-edta tetrahydrate, copper ii disodium 2,2',2,2'-ethane-1,2-diylbis azanetriyl tetraacetate tetrahydrate
PubChem CID 46245299
IUPAC Name copper;disodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;tetrahydrate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.O.O.O.[Na+].[Na+].[Cu+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E009025G
View Documents
TCI America
E009025G
25 g
Each for $57.79
Only null left
 
E0090500G
View Documents
TCI America
E0090500G
500 g
Each for $337.40
Only null left
 
Description

Description

Specifications

Chemical Identifiers

39208-15-6
469.798
CHPMNDHAIUIBSK-UHFFFAOYSA-J
46245299
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.O.O.O.[Na+].[Na+].[Cu+2]
C10H20CuN2Na2O12
MFCD00148920
copper ii disodium ethylenediaminetetraacetate tetrahydrate, ethylenediaminetetraacetic acidcopper ii disodium salt tetrahydrate, acmc-209j3w, edta-cu-15, ksc911m0f, copper 2+ disodium ion 4-edta tetrahydrate, copper ii disodium 2,2',2,2'-ethane-1,2-diylbis azanetriyl tetraacetate tetrahydrate
copper;disodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;tetrahydrate

Specifications

39208-15-6
C10H20CuN2Na2O12
25 g
CHPMNDHAIUIBSK-UHFFFAOYSA-J
copper;disodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;tetrahydrate
46245299
≥98.0% (T)
Copper(II) Disodium Ethylenediaminetetraacetate Tetrahydrate
Blue
MFCD00148920
copper ii disodium ethylenediaminetetraacetate tetrahydrate, ethylenediaminetetraacetic acidcopper ii disodium salt tetrahydrate, acmc-209j3w, edta-cu-15, ksc911m0f, copper 2+ disodium ion 4-edta tetrahydrate, copper ii disodium 2,2',2,2'-ethane-1,2-diylbis azanetriyl tetraacetate tetrahydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.O.O.O.[Na+].[Na+].[Cu+2]
469.798
397.74
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1266

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'