accessibility menu, dialog, popup

UserName

NOOOONOOOOH2OH2OH2OH2ONaNaCu

CAS RN 39208-15-6

IUPAC Name: copper(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate tetrahydrate
Synonyms: copper(2+) disodium ion(4-) edta tetrahydrate
Molecular Weight (g/mol): 469.8
Molecular Formula: C10H20CuN2Na2O12
InChi Key: CHPMNDHAIUIBSK-UHFFFAOYSA-J
SMILES: O.O.O.O.[Na+].[Na+].[Cu++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results