missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Disodium Ethylenediaminetetraacetate Tetrahydrate 98.0+%, TCI America™

Catalog No. E0090500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0090500G 500 g
E009025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0090500G Supplier TCI America Supplier No. E0090500G
Only null left

Chemical Identifiers

CAS 39208-15-6
Molecular Formula C10H20CuN2Na2O12
Molecular Weight (g/mol) 469.798
MDL Number MFCD00148920
InChI Key CHPMNDHAIUIBSK-UHFFFAOYSA-J
Synonym copper ii disodium ethylenediaminetetraacetate tetrahydrate, ethylenediaminetetraacetic acidcopper ii disodium salt tetrahydrate, acmc-209j3w, edta-cu-15, ksc911m0f, copper 2+ disodium ion 4-edta tetrahydrate, copper ii disodium 2,2',2,2'-ethane-1,2-diylbis azanetriyl tetraacetate tetrahydrate
PubChem CID 46245299
IUPAC Name copper;disodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;tetrahydrate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.O.O.O.[Na+].[Na+].[Cu+2]

Specifications

Color Blue
Quantity 500 g
Formula Weight 397.74
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Copper(II) Disodium Ethylenediaminetetraacetate Tetrahydrate
EINECSNumber (2)-1266
TSCA No