missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) Disodium Ethylenediaminetetraacetate Tetrahydrate 98.0+%, TCI America™
Specifications
Specifications
| Color | Blue |
| Quantity | 25 g |
| Formula Weight | 397.74 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Copper(II) Disodium Ethylenediaminetetraacetate Tetrahydrate |
Safety and Handling
| EINECSNumber | (2)-1266 |
| TSCA | No |