missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Diethyldithiocarbamate 97.0+%, TCI America™

Catalog No. p-7126242
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D04875G 5 g
D048725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D04875G Supplier TCI America Supplier No. D04875G

Chemical Identifiers

CAS 13681-87-3
Molecular Formula C10H20CuN2S4
Molecular Weight (g/mol) 360.07
MDL Number MFCD00070506
InChI Key OBBCYCYCTJQCCK-UHFFFAOYSA-L
Synonym Diethyldithiocarbamic Acid Copper(II) Salt
PubChem CID 26180
IUPAC Name copper;N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)[S-].CCN(CC)C(=S)[S-].[Cu+2]

Specifications

Melting Point 197°C
Quantity 5 g
Formula Weight 360.07
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Copper(II) Diethyldithiocarbamate
RTECSNumber EZ5250000
TSCA No