accessibility menu, dialog, popup

UserName

CuH3CNH3CSSH3CNCH3SS

CAS RN 13681-87-3

IUPAC Name: copper(2+) bis((diethylcarbamothioyl)sulfanide)
Synonyms: copper(2+) didiethyldithiocarbamate
Molecular Weight (g/mol): 360.07
Molecular Formula: C10H20CuN2S4
InChi Key: OBBCYCYCTJQCCK-UHFFFAOYSA-L
SMILES: [Cu++].CCN(CC)C([S-])=S.CCN(CC)C([S-])=S
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results