missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) bromide, 99%

CAS: 7789-45-9 | CuBr2

Supplier:  Thermo Scientific Chemicals A1633514

 View more versions of this product

Catalog No. AAA1633514


Only null left
Explore available promotions

Description

Description

Copper(II) bromide is used in photographic processing as an intensifier and as a brominating agent in organic synthesis. It is also used in the coupling of o-iodophenols and aryl acetylenes avoiding the use of palladium. It finds application in the preparation of NK1/NK2 receptor antagonists used in the regulation of tachykinin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) bromide is used in photographic processing as an intensifier and as a brominating agent in organic synthesis. It is also used in the coupling of o-iodophenols and aryl acetylenes avoiding the use of palladium. It finds application in the preparation of NK1/NK2 receptor antagonists used in the regulation of tachykinin.

Solubility
Soluble in water, alcohol, ammonia and acetone. Insoluble in benzene, ether, sulfuric acid and ethyl ether.

Notes
Hygroscopic. Incompatible with oxidizing agents and metals.
Specifications

Specifications

900°C
498°C
25 g
MFCD00010970
14,2629
Soluble in water,alcohol,ammonia and acetone. Insoluble in benzene,ether,sulfuric acid and ethyl ether.
99%
Hygroscopic
Copper(II) bromide
7789-45-9
CuBr2
UN3260
Cupric bromide
223.36
Odorless
4.71

Chemical Identifiers

7789-45-9
CuBr2
MFCD00010970
Cupric bromide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 232-167-2

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only