missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) bromide, 99+%, extra pure, anhydrous

Catalog No. AC206331000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC206331000 100 g Glass bottle
AC206330050 5 g Glass bottle
AC206335000 500 g Glass bottle
AC206330025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC206331000 Supplier Thermo Scientific Chemicals Supplier No. 206331000

CAS: 7789-45-9 | Br2Cu | 223.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 7789-45-9
Molecular Formula Br2Cu
Molecular Weight (g/mol) 223.36
MDL Number MFCD00010970
Synonym Cupric bromide

Specifications

Boiling Point 900.0°C
Melting Point 498.0°C
Color Black-Gray to Gray
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99% min. (Iodometry)
Linear Formula CuBr2
Fieser 01,161; 02,84; 05,158; 06,138; 13,112; 15,100
Merck Index 15, 2619
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, acetone, nh3, practically insoluble in benzene, diethyl ether,, conc. h2so4
Formula Weight 223.36
Percent Purity ≥99%
Grade Extra Pure
Packaging Glass bottle
Density 4.77
Chemical Name or Material Copper(II) bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 232-167-2
TSCA TSCA

RUO – Research Use Only