missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) bromide, 99%, for analysis, anhydrous

Catalog No. p-7077451
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453952500 250 g
AC453950010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453952500 Supplier Thermo Scientific Chemicals Supplier No. 453952500
Only null left

CAS: 7789-45-9 | Br2Cu | 223.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-45-9
Molecular Formula Br2Cu
Molecular Weight (g/mol) 223.36
Synonym Cupric bromide

Specifications

Boiling Point 900.0°C
Melting Point 498.0°C
Quantity 250 g
Assay Percent Range 98.5% min. (Iodometry)
Linear Formula CuBr2
Merck Index 15,2619
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, acetone, nh3, practically insoluble in benzene, diethyl ether,, conc. h2so4
Formula Weight 223.36
Percent Purity 99%
Grade Analytical
Loss on Drying 1% max.
Chemical Name or Material Copper(II) bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 232-167-2

RUO – Research Use Only