Learn More
Copper(I) telluride, 99.5% (metals basis)
CAS: 12019-52-2 | Cu2Te | 254.692 g/mol
$134.88 - $552.53
Chemical Identifiers
| CAS | 12019-52-2 |
|---|---|
| Molecular Formula | Cu2Te |
| Molecular Weight (g/mol) | 254.692 |
| MDL Number | MFCD00049727 |
| InChI Key | MZEWONGNQNXVKA-UHFFFAOYSA-N |
| Synonym | copper telluride, cuprous telluride, copper i telluride, dicopper telluride, cuprous monotelluride, dicopper monotelluride, copper telluride cu0.67te0.33, cupriotellanyl copper |
| PubChem CID | 6914517 |
| IUPAC Name | copper;tellurium |
| SMILES | [Cu].[Cu].[Te] |
Description
It is used as a colorent of glass and ceramics, rubber vulcanization accelerator, metallurgical additives, petrochemical catalyst, solar cells, thermoelectric refrigeration Materials, Infrared Detection Materials, Electroplating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12019-52-2 | |
| 254.692 | |
| MZEWONGNQNXVKA-UHFFFAOYSA-N | |
| 6914517 | |
| [Cu].[Cu].[Te] |
| Cu2Te | |
| MFCD00049727 | |
| copper telluride, cuprous telluride, copper i telluride, dicopper telluride, cuprous monotelluride, dicopper monotelluride, copper telluride cu0.67te0.33, cupriotellanyl copper | |
| copper;tellurium |
Specifications
| Black-Blue | |
| Pieces | |
| 99.5% (Metals basis) | |
| MFCD00049727 | |
| Insoluble in water. | |
| [Cu].[Cu].[Te] | |
| 254.692 | |
| 254.68 | |
| 1.45 | |
| Copper(I) telluride |
| 12019-52-2 | |
| 5 g | |
| Cu2Te | |
| copper telluride, cuprous telluride, copper i telluride, dicopper telluride, cuprous monotelluride, dicopper monotelluride, copper telluride cu0.67te0.33, cupriotellanyl copper | |
| MZEWONGNQNXVKA-UHFFFAOYSA-N | |
| copper;tellurium | |
| 6914517 | |
| Odorless | |
| 7.27 g/cm3 |
Safety and Handling
Keep container in a well ventilated place. Wear suitable protective clothing. This material and/or its container must be disposed of as hazardous waste.
H301-H332; oxic if swallowed. Harmful if inhaled.
EINECSNumber : 234-646-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only