missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) telluride, 99.5% (metals basis)

Catalog No. AA1314006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1314006 5 g
AA1314014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1314006 Supplier Thermo Scientific Chemicals Supplier No. 01314006
Only null left

CAS: 12019-52-2 | Cu2Te | 254.692 g/mol

It is used as a colorent of glass and ceramics, rubber vulcanization accelerator, metallurgical additives, petrochemical catalyst, solar cells, thermoelectric refrigeration Materials, Infrared Detection Materials, Electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12019-52-2
Molecular Formula Cu2Te
Molecular Weight (g/mol) 254.692
MDL Number MFCD00049727
InChI Key MZEWONGNQNXVKA-UHFFFAOYSA-N
Synonym copper telluride, cuprous telluride, copper i telluride, dicopper telluride, cuprous monotelluride, dicopper monotelluride, copper telluride cu0.67te0.33, cupriotellanyl copper
PubChem CID 6914517
IUPAC Name copper;tellurium
SMILES [Cu].[Cu].[Te]

Specifications

Color Black-Blue
Physical Form Pieces
Quantity 5 g
Assay Percent Range 99.5% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 254.68
Odor Odorless
Refractive Index 1.45
Density 7.27 g/cm3
Chemical Name or Material Copper(I) telluride
Hazard Category H301-H332; oxic if swallowed. Harmful if inhaled.
Hazard Statement Keep container in a well ventilated place. Wear suitable protective clothing. This material and/or its container must be disposed of as hazardous waste.
EINECSNumber 234-646-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only