missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Oxide, Honeywell Fluka™

Catalog No. p-7071790
Change view
Click to view available options
Quantity:
2.5 kg
6 x 2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000435 2.5 kg
60044764 6 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000435 Supplier Honeywell Chemicals Supplier No. 128412.5KG
Only null left

≥86% Cu basis, red

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1317-39-1
Molecular Formula Cu2O
MDL Number MFCD00010974

Specifications

Color Red
Quantity 2.5 kg
UN Number 3077
Formula Weight 143.09g/mol
Percent Purity ≥86% (Cu basis)
Packaging Plastic Bottle
Chemical Name or Material Copper(I) Oxide
Hazard Statement P261-P301 + P312 + P330-P305 + P351 + P338
Precautionary Statement H302 + H332-H319-H410
EINECSNumber 215-270-7