accessibility menu, dialog, popup

UserName

H2OCuCu

CAS RN 1317-39-1

IUPAC Name: bis(λ¹-copper(1+)) oxidandiuide
Synonyms: cuprous oxide
Molecular Weight (g/mol): 145.11
Molecular Formula: Cu2H2O
InChi Key: FDARDWGARJUOKS-UHFFFAOYSA-N
SMILES: [OH2--].[Cu+].[Cu+]
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (5)
  • (1)
Physical Form 
  • (13)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (2)
  • (7)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (13)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (5)
  • (1)
  • (13)
  • (3)
  • (3)
  • (6)
  • (2)
  • (7)
  • (3)