missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(I) oxide, 99.9% (metals basis)
Description
It is used chiefly as a pigment in ceramics and in antifouling paints.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1800°C (decomposition) |
| Melting Point | 1235°C |
| Color | Purple-Red |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 99% (Metal basis) |
| Solubility Information | Insoluble in water but soluble in acids. |
| Formula Weight | 143.08 |
| Odor | Odorless |
| Refractive Index | 2.705 |
| Show More |
Safety and Handling
| Hazard Category | H302+H332-H318 |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P310-P312-P330-P501c |
| DOTInformation | DOT Class: 9, Packing Group: III |
| EINECSNumber | 215-270-7 |
| RTECSNumber | GL8050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only