missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) oxide, 97%

Catalog No. p-46548
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC197425000 500 g Glass bottle
AC197420025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC197425000 Supplier Thermo Scientific Chemicals Supplier No. 197425000
Only null left

CAS: 1317-39-1 | Cu2O | 143.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1317-39-1
Molecular Formula Cu2O
Molecular Weight (g/mol) 143.09
MDL Number MFCD00010974
Synonym Cuprous oxide

Specifications

Boiling Point 1800.0°C
Melting Point 1232.0°C
Color Brown-Red
Physical Form Powder
Quantity 500 g
Assay Percent Range 96% min. (Complexometry)
Linear Formula Cu2O
Merck Index 15, 2654
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ammonia or min.acids
Formula Weight 143.09
Percent Purity 97%
Packaging Glass bottle
Chemical Name or Material Copper(I) oxide, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 215-270-7
RTECSNumber GL8050000
TSCA TSCA

RUO – Research Use Only