missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) oxide, 97%

Catalog No. AAA1443622
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1443622 100 g
AAA1443636 500 g
AAA144360E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1443622 Supplier Thermo Scientific Chemicals Supplier No. A1443622
Only null left

CAS: 1317-39-1 | Cu2O

It is used as a catalyst in decarboxylation reactions of arylacetic acids and of malonic acids under mild conditions. It is also used in batteries and electrodes, ceramic colorant, catalyst, purification of hydrogen, reagent in analytical chemistry, metallurgical and welding fluxes. It finds its application also as an antifouling agent for marine paints and in Benedict's Test. It is used to provide a source supplemental elemental copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a catalyst in decarboxylation reactions of arylacetic acids and of malonic acids under mild conditions. It is also used in batteries and electrodes, ceramic colorant, catalyst, purification of hydrogen, reagent in analytical chemistry, metallurgical and welding fluxes. It finds its application also as an antifouling agent for marine paints and in Benedict′s Test. It is used to provide a source supplemental elemental copper.

Solubility
Insoluble in water. Dissolves in concentrated ammonia.

Notes
Air and moisture sensitive. Store away from air, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1317-39-1
Molecular Formula Cu2O
MDL Number MFCD00010974
Synonym Cuprous oxide

Specifications

Boiling Point 1800°C (decomposition)
Melting Point 1235°C
Quantity 100 g
UN Number UN3077
Merck Index 14,2664
Solubility Information Insoluble in water. Dissolves in concentrated ammonia.
Formula Weight 143.08
Percent Purity 97%
Odor Odorless
Sensitivity Air and moisture sensitive
Density 6
Chemical Name or Material Copper(I) oxide
Show More Show Less
Hazard Category H302+H332-H318
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P310-P312-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 215-270-7
RTECSNumber GL8050000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only