Learn More
Copper(I) oxide, 97% (Cu + Cu{2}O Assay)
Description
Red pigment for glass and ceramic glazes, and as a catalystCopper(I) oxide is used in antifouling, marine paint, brazing paste, antifungal treatment for plant seeds. Used as a hydrogen scavenger in copper brass and bronze and high grade lubricants. Finds uses in glass and ceramic pigments, ferrite magnets, catalyst in organic synthesis, dyes and dye intermediates and as catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1800°C (decomposition) |
| Melting Point | 1235°C |
| Color | Purple-Red |
| Physical Form | Powder |
| Quantity | 500 g |
| Assay Percent Range | 97% |
| Solubility Information | Practically insoluble in water. Soluble in NH4OHInsoluble in water but soluble in acids. |
| Formula Weight | 143.08 |
| Odor | Odorless |
| Refractive Index | 2.705 |
| Show More |
Safety and Handling
| Hazard Category | H302+H332-H318 |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P310-P312-P330-P501c |
| DOTInformation | DOT Class: 9, Packaging Group: III |
| EINECSNumber | 215-270-7 |
| RTECSNumber | GL8050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only