missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) oxide, 97% (Cu + Cu{2}O Assay)

Catalog No. AA1230036
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1230036 500 g
AA12300A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1230036 Supplier Thermo Scientific Chemicals Supplier No. 01230036

Red Pigment For Glass and Ceramic Glazes, and As a Catalyst | CAS: 1317-39-1 | Cu2O

Red pigment for glass and ceramic glazes, and as a catalystCopper(I) oxide is used in antifouling, marine paint, brazing paste, antifungal treatment for plant seeds. Used as a hydrogen scavenger in copper brass and bronze and high grade lubricants. Finds uses in glass and ceramic pigments, ferrite magnets, catalyst in organic synthesis, dyes and dye intermediates and as catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1317-39-1
Molecular Formula Cu2O
MDL Number MFCD00010974

Specifications

Boiling Point 1800°C (decomposition)
Melting Point 1235°C
Color Purple-Red
Physical Form Powder
Quantity 500 g
Assay Percent Range 97%
Solubility Information Practically insoluble in water. Soluble in NH4OHInsoluble in water but soluble in acids.
Formula Weight 143.08
Odor Odorless
Refractive Index 2.705
Sensitivity Air and moisture sensitive
Appearance Purple-red powder
Density 6 g/cm3
Chemical Name or Material Copper(I) oxide, Cu + Cu2O Assay
Show More Show Less
Hazard Category H302+H332-H318
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P310-P312-P330-P501c
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 215-270-7
RTECSNumber GL8050000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only