missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) bromide, 98%, extra pure

Catalog No. p-46506
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC199582500 250 g Glass bottle
AC199580010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC199582500 Supplier Thermo Scientific Chemicals Supplier No. 199582500
Only null left

CAS: 7787-70-4 | BrCu | 143.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7787-70-4
Molecular Formula BrCu
Molecular Weight (g/mol) 143.45
MDL Number MFCD00010969
InChI Key NKNDPYCGAZPOFS-UHFFFAOYSA-M
Synonym copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide
PubChem CID 24593
IUPAC Name bromocopper
SMILES [Cu+].[Br-]

Specifications

Boiling Point 1345.0°C
Melting Point 504.0°C
Color Green
Physical Form Powder
Quantity 250 g
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula CuBr
Fieser 01,165; 02,90; 03,67; 04,108; 05,163; 06,143; 07,79; 08,116; 11,140; 12,286; 14,99; 16,96
Merck Index 15, 2649
Solubility Information Solubility in water: soluble in cold water. Other solubilities: decomposes in hot water, sol. in hcl, hbr, nh4oh (formation of complexes), prectically insoluble in conc. h2so4 and acetone, insoluble matter in hcl: ≤0.02%
Formula Weight 143.45
Percent Purity 98%
Grade Extra Pure
Packaging Glass bottle
Specific Gravity 4.71
Density 4.7100g/mL
Chemical Name or Material Copper(I) bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 232-131-6
TSCA TSCA

RUO – Research Use Only