missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Acetate 93.0+%, TCI America™

Catalog No. p-7117503
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15405G 5 g
A154025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15405G Supplier TCI America Supplier No. A15405G

Chemical Identifiers

CAS 598-54-9
Molecular Formula C2H3CuO2
Molecular Weight (g/mol) 122.59
MDL Number MFCD00058908
InChI Key RFKZUAOAYVHBOY-UHFFFAOYSA-M
Synonym copper i acetate, cuprous acetate, copper 1+ acetate, acetic acid, copper 1+ salt, unii-9u3n93lmzb, copper acetate, 9u3n93lmzb, acetic acid, copper 1+ salt 1:1, copper 1 acetate
PubChem CID 69023
IUPAC Name copper(1+);acetate
SMILES CC(=O)[O-].[Cu+]

Specifications

Quantity 5 g
Formula Weight 122.59
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Copper(I) Acetate
EINECSNumber (2)-0693
TSCA Yes