accessibility menu, dialog, popup

UserName

CuCH3OO

CAS RN 598-54-9

IUPAC Name: λ¹-copper(1+) acetate
Synonyms: λ¹-copper(1+) acetate
Molecular Weight (g/mol): 122.59
Molecular Formula: C2H3CuO2
InChi Key: RFKZUAOAYVHBOY-UHFFFAOYSA-M
SMILES: [Cu+].CC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results