missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(I) Acetate 93.0+%, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| Formula Weight | 122.59 |
| Percent Purity | ≥93.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Copper(I) Acetate |
Safety and Handling
| EINECSNumber | (2)-0693 |
| TSCA | Yes |