Learn More
Copper aluminum oxide, 99.5% (metals basis)
CAS: 12042-92-1 | Al2CuO4 | 181.51 g/mol
Supplier: Thermo Scientific Chemicals 03941918
Description
Applications for copper aluminum oxide nanocrystalsare included in microbatteries, in micro-electronics, circuits and in nanowire, and in nanofluids.It is also a promising material for thermoelectric devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Copper aluminum oxide | |
| Powder | |
| (metals basis) | |
| MFCD00210610 | |
| IPJRADJOSNLGBF-UHFFFAOYSA-N | |
| copper;oxido(oxo)alumane | |
| 12947132 | |
| 99.5% |
| 12042-92-1 | |
| 50 g | |
| Al2CuO4 | |
| Insoluble in water. | |
| [Cu++].[O-][Al]=O.[O-][Al]=O | |
| 181.51 | |
| 181.5 |
Chemical Identifiers
| 12042-92-1 | |
| 181.51 | |
| IPJRADJOSNLGBF-UHFFFAOYSA-N | |
| copper;oxido(oxo)alumane |
| Al2CuO4 | |
| MFCD00210610 | |
| 12947132 | |
| [Cu++].[O-][Al]=O.[O-][Al]=O |
Safety and Handling
EINECSNumber : 234-934-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only