missing translation for 'onlineSavingsMsg'
Learn More

Copper aluminum oxide, 99.5% (metals basis)

CAS: 12042-92-1 | Al2CuO4 | 181.51 g/mol

$229.49 - $229.49

Chemical Identifiers

CAS 12042-92-1
Molecular Formula Al2CuO4
Molecular Weight (g/mol) 181.51
MDL Number MFCD00210610
InChI Key IPJRADJOSNLGBF-UHFFFAOYSA-N
PubChem CID 12947132
IUPAC Name copper;oxido(oxo)alumane
SMILES [Cu++].[O-][Al]=O.[O-][Al]=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3941918
View Documents
Thermo Scientific Chemicals
03941918
50 g
Each for $229.49
Only null left
 
Description

Description

Applications for copper aluminum oxide nanocrystalsare included in microbatteries, in micro-electronics, circuits and in nanowire, and in nanofluids.It is also a promising material for thermoelectric devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12042-92-1
181.51
IPJRADJOSNLGBF-UHFFFAOYSA-N
copper;oxido(oxo)alumane
Al2CuO4
MFCD00210610
12947132
[Cu++].[O-][Al]=O.[O-][Al]=O

Specifications

12042-92-1
50 g
MFCD00210610
IPJRADJOSNLGBF-UHFFFAOYSA-N
copper;oxido(oxo)alumane
12947132
99.5%
(metals basis)
Powder
Al2CuO4
Insoluble in water.
[Cu++].[O-][Al]=O.[O-][Al]=O
181.51
181.5
Copper aluminum oxide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 234-934-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only