missing translation for 'onlineSavingsMsg'
Learn More

Copper, 99+%, turnings

CAS: 7440-50-8 | Cu | 63.55 g/mol

Supplier:  Thermo Scientific Chemicals 317910010

 View more versions of this product

Catalog No. AC317910010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Copper
1083.0°C
2595.0°C
1 kg
Turnings
Cu
MFCD00010965
15, 2504
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
23978
63.54
99+%
7440-50-8
8.92
Glass bottle
99+%
Cu
01,157; 02,82; 03,63; 04,102; 05,146; 07,73; 08,113; 09,122; 15,99; 16,95
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
63.55
CHEBI:30052
≥99%

Chemical Identifiers

7440-50-8
63.55
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

MOISTURE SENSITIVE

EINECSNumber : 231-159-6

RTECSNumber : GL5325000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only