missing translation for 'onlineSavingsMsg'
Learn More

Copper, 99+%, turnings

Catalog No. p-222929
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC317912500 250 g Glass bottle
AC317910010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC317912500 Supplier Thermo Scientific Chemicals Supplier No. 317912500
Only null left

CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Melting Point 1083.0°C
Boiling Point 2595.0°C
Density 8.92
Quantity 250 g
Packaging Glass bottle
Form Turnings
Assay Percent Range 99+%
Linear Formula Cu
Fieser 01,157; 02,82; 03,63; 04,102; 05,146; 07,73; 08,113; 09,122; 15,99; 16,95
Merck Index 15, 2504
Formula Weight 63.54
Percent Purity ≥99%
Chemical Name or Material Copper, 99+%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
EINECSNumber 231-159-6
RTECSNumber GL5325000
TSCA TSCA

RUO – Research Use Only