missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Copolyester, Carboy
For Use With: For storing and dispensing solvents, buffers, reagents and powders. Especially when visual inspection needed.; Closure Material: Polypropylene; Color: Clear
$269.22 - $469.63
Specifications
| Accuracy | ±5% |
|---|---|
| Closure Material | Polypropylene |
| Closure Size | 83 mm |
| Color | Clear |
| For Use With (Application) | For storing and dispensing solvents, buffers, reagents and powders. Especially when visual inspection needed. |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|
19-830-446
|
Justrite™
12946 |
2.5 L |
Each for $269.22
|
|
|||||
|
19-830-447
|
Justrite™
12947 |
5 L |
Each for $369.95
|
|
|||||
|
19-830-448
|
Justrite™
JMC12948 |
10 L |
Each for $469.63
|
|
|||||
Description
- Constructed from clear, BPA-free, USP Class VI, FDA grade resin
- Rectangular ergonomic design
- Interchangeable caps and adapters provide top connection flexibility
- Material, volume, and metric graduation marks imprinted on ca
Specifications
| ±5% | |
| 83 mm | |
| For storing and dispensing solvents, buffers, reagents and powders. Especially when visual inspection needed. | |
| Carboy |
| Polypropylene | |
| Clear | |
| Copolyester | |
| Rectangular |