missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Copolyester, Carboy
GREENER_CHOICE

Catalog No. 19830446
Change view
Click to view available options
Capacity (Metric):
2.5 L
5 L
10 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
19830446 2.5 L
19830447 5 L
19830448 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19830446 Supplier Justrite™ Supplier No. 12946

For Use With: For storing and dispensing solvents, buffers, reagents and powders. Especially when visual inspection needed.; Closure Material: Polypropylene; Color: Clear

  • Constructed from clear, BPA-free, USP Class VI, FDA grade resin
  • Rectangular ergonomic design
  • Interchangeable caps and adapters provide top connection flexibility
  • Material, volume, and metric graduation marks imprinted on ca

Specifications

Accuracy ±5%
Capacity (English) 0.7 gal.
Capacity (Metric) 2.5 L
Closure Material Polypropylene
Closure Size 83 mm
Color Clear
Dimensions (L x W x H) 170 x 127 x 205 mm
For Use With (Application) For storing and dispensing solvents, buffers, reagents and powders. Especially when visual inspection needed.
Height (English) 12 in.
Length (English) 6.7 in.
Material Copolyester
Product Type Carboy
Shape Rectangular
Width (English) 5 in.
Show More Show Less