missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COPII Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1069A
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1069A 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1069A Supplier Invitrogen™ Supplier No. PA1069A
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

PA1-069A detects coatomer-protein II (COPII) from rat, mouse and human cells. PA1-069A has been successfully used in Western blot and ICC/IF procedures. By Western blot, this antibody detects an ~85 kDa protein representing COPII from PC-12 cell extract. The PA1-069A immunogen is a synthetic peptide corresponding to residues M(1) T T Y L E F I Q Q N E E R D G V R (18) C of rat COPII. This sequence is completely conserved between human, mouse, and rat. PA1-069A immunizing peptide (Cat. # PEP-127) is available for use in neutralization and control experiments.

Coatomer proteins are involved in regulating transport between the endoplasmic reticulum (ER) and the Golgi complex and in intra-Golgi transport. There exist two coatomer-protein mechanisms (COPI and COPII) and although they have mechanistic parallels, they are molecularly distinct. The COPI coat is comprised of seven subunits (alpha-, beta-, beta'-, gamma-, delta-, epsilon-, and zeta-COP) in a complex called coatomer. Assembly of the coatomer (COPI) onto non-clathrin coated vesicles is regulated by ADP-ribosylation factor (ARF). Vesicle formation, budding, fusion, and disassembly is dependent on GDP-GTP exchange, COPI, and ARF. COPI has been shown to facilitate retrograde intracellular transport from the ER to the Golgi complex. By contrast, COPII facilitates anterograde transport between these subcellular organelles. COPII has been shown to be independently and selectively recruited to the ER relative to COPI subunits. COPII consists of three parts: Sar1p and the two protein complexes, Sec23p-Sec24p and Sec13p-Sec31p.

Specifications

Antigen COPII
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene SEC24A
Gene Accession No. O94979, O95486, P55735, Q01405, Q15436, Q3U2P1, Q3UPL0, Q5XFW8, Q9D1M0, Q9Z2Q1
Gene Alias 1110003H02Rik; 1810024J13Rik; ABP125; ABP130; AU042160; CLSD; cop ii; D3S1231E; GATOR complex protein SEC13; hSec23A; HSPC275; HSPC334; KIAA0905; Msec23; npp-20; Protein SEC13 homolog; protein transport protein Sec23A; Protein transport protein Sec24A; protein transport protein Sec31A; Sec13; SEC13 homolog; SEC13 homolog (S. cerevisiae); SEC13 homolog, nuclear pore and COPII coat complex component; SEC13 homolog, nuclear pore and COPII coating complex component; SEC13 related; SEC13A; SEC13L1; SEC13-like 1; SEC13-like 1 isoform; SEC13-like protein 1; Sec13r; SEC13-related protein; Sec23; Sec23 homolog A (S. cerevisiae); Sec23 homolog A, coat complex II component; Sec23 homolog A, COPII coat complex component; SEC23A; SEC23A (S. cerevisiae); Sec23r; SEC23-related protein A; SEC24 family member A; SEC24 family, member A; SEC24 homolog A, COPII coat complex component; SEC24 related gene family, member A; Sec24 related gene family, member A (S. cerevisiae); SEC24A; SEC24-related protein A; SEC31 homolog A (S. cerevisiae); SEC31 homolog A, COPII coat complex component; SEC31 homolog A, COPII coating complex component; Sec31a; SEC31L1; SEC31-like 1; SEC31-like protein 1; SEC31-related protein A; VAP1; vesicle associated protein; vesicle-associated protein 1; Web1-like protein; yeast Sec31p homolog
Gene Symbols SEC13, SEC23A, SEC24A, SEC31A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues M(1) T T Y L E F I Q Q N E E R D G V R (18) C of rat COPII.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10484, 10802, 110379, 20334, 22872, 287275, 297522, 58817, 6396, 69162, 77371, 93646
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less