Learn More
Copan Diagnostics T-Shaped Spreaders
T-shaped spreaders
$256.12
Specifications
| Material | Plastic |
|---|---|
| Product Type | Cell Spreader |
| Sterility | Sterile |
| For Use With (Equipment) | Culture Plates |
| Shape | T-Shape |
Description
T-shaped spreaders
T-Shaped Plastic Spreaders
- Designed for spreading and dispersing liquids across the surface of agar culture plates
- Used for performing bacterial CFU plate counts on water, milk, urine and other liquid samples
- Smooth, rounded surfaces prevent any cutting or gouging of agar during inoculation of culture plates
- T-Shape design means that even pressure is applied across the entire length of the T-bar in contact with the agar
- Available individually sterile wrapped or in packs of five or ten pieces in a peel pack
Specifications
| Plastic | |
| Sterile | |
| T-Shape |
| Cell Spreader | |
| Culture Plates | |
| Designed for spreading and dispersing liquids across the surface of agar culture plates, used for performing bacterial CFU plate counts on water, milk, urine and other liquid samples |