Learn More
CoorsTek™ Porcelain Casserole Evaporating Dishes
Withstand temperatures to 1150°C (2102°F)
| Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||
|
04695F
|
CoorsTek™
CRS66060 |
210 mL | 100 mm | 50 mm |
N/A
|
|
|||
|
04695K
|
CoorsTek™
CRS66064/DUP |
1200 mL | 174 mm | 100 mm |
N/A
|
|
|||
Description
- Glazed inside and outside except on rims
- With lip
Specifications
| Porcelain | |
| Glazed |